Learn More
Merck Millicell™ Receiver Trays
Comes with lid
£161.00 - £165.00
Specifications
Closure Included With Lid
Color Clear
Material Polystyrene
Shape Rectangular
Sterility Sterile
View More Specs
Products 2
Product Code Brand Well Count Price Quantity & Availability  
Product Code Brand Well Count Price Quantity & Availability  
10347753
View Documents
Merck
PSSW010R5
1
£161.00
Pack of 5
Estimated Shipment: 29-05-2025
to see stock.
Add to basket
10052523
View Documents
Merck
PSMW010R5
24
£165.00
Each
Estimated Shipment: 25-06-2025
to see stock.
Add to basket
Description
Description
Designed to support cell attachment, growth, and differentiation for a variety of applications
  • 24-well feeder tray incorporates a tear-drop well design to reduce the incidence of air bubbles that can form under the membrane during plate assembly, ensuring that cell monolayers are getting the proper nutrients
  • Single-well feeder tray utilizes baffles to reduce media leakage and contamination
Specifications
Specifications
With Lid
Polystyrene
Sterile
Clear
Rectangular
Receiver Tray
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Merck Millicell™ Receiver Trays