Learn More
Merck Millicell™ Receiver Trays
Comes with lid
Brand:  Merck PSMW010R5
 View more versions of this product
Product Code. 10052523
  • £165.00 / Each
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Designed to support cell attachment, growth, and differentiation for a variety of applications
  • 24-well feeder tray incorporates a tear-drop well design to reduce the incidence of air bubbles that can form under the membrane during plate assembly, ensuring that cell monolayers are getting the proper nutrients
  • Single-well feeder tray utilizes baffles to reduce media leakage and contamination
TRUSTED_SUSTAINABILITY
Specifications
Specifications
With Lid
Polystyrene
Sterile
24
PCF
Clear
Rectangular
Receiver Tray
24-Well Receiver Tray, Lid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Merck Millicell™ Receiver Trays > Cell Culture Microplate; 24-well Receiver Plate and Lid