missing translation for 'onlineSavingsMsg'
Learn More
Mineral oil, pure, Thermo Scientific Chemicals Product Code.: 10219650

Mineral oil, pure, Thermo Scientific Chemicals

Product Code. 10219650
1 L
Click to view available options
Quantity:
6 mL
1 L
2.5 L
10 L
This item is not returnable. View return policy

Product Code. 10219650

Brand: Thermo Scientific Acros 415080010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8042-47-5
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.363
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
Synonym acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g
PubChem CID 9566064
IUPAC Name disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color Colorless
Density 0.8770g/mL
Flash Point 220°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4760 to 1.4830
Quantity 1 L
Specific Gravity 0.877
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene, chloroform, ether, cs2,, petroleum ether, oils, miscible with wax and fats, insoluble in ethanol
Viscosity 65-75 mm2/s (40°C)
Grade Pure
Physical Form Oily Liquid
Chemical Name or Material Mineral oil
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465644 Certificate TypeCertificate of Analysis Date28/08/2024 Catalog Number415080010,415080060,415080025,415080100
Lot NumberA0465645 Certificate TypeCertificate of Analysis Date26/08/2024 Catalog Number415080010,415080060,415080025,415080100
Lot NumberA0459751 Certificate TypeCertificate of Analysis Date30/01/2024 Catalog Number415080010,415080060,415080025,415080100
Lot NumberA0423138 Certificate TypeCertificate of Analysis Date11/09/2023 Catalog Number415080010,415080060,415080025,415080100
Lot NumberA0453791 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number415080010,415080060,415080025,415080100

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mineral oil, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.