Learn More
3M™ Mini Flip-Top Vial
Encompass
Leak-resistant plastic cylinder with a flip-open lid that seals securely for safe shipping and handling.
Encompass_Preferred
POA - £109.26
Specifications
Quantity 100/Cs.
View More Specs
Products 3
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
17298520
Encompass_Preferred
View Documents
3M™
7000030317
9 mL Maximum Recovery Diluent
£109.26
Pack of 100
View alternative products
Save Up to 
»
17223734
Encompass
View Documents
3M™
7000053825
9 mL Letheen Broth
POA
Pack of 100
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
17263734
Encompass
View Documents
3M™
7000053834
Buffered Peptone Water Broth
POA
Pack of 100
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • Hinged lid can be opened with one hand and stays open for easy sample additions.
  • Decontaminated through gamma irradiation and ready for use with no additional blending required.
  • Flat bottom tube stands on its own, no holder required.
  • Leak resistant vial helps prevent leakage during shipping.
  • 2 year shelf life from date of manufacture provides long-term readiness.
Specifications
Specifications
100/Cs.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
3M™ Mini Flip-Top Vial