Learn More
Cytiva (formerly Pall Lab) Mini Profile™ Capsules with Profile II Filter
Mini Profile™ Capsules designed for research applications only.
Brand:  Cytiva (formerly Pall Lab) 12073
Additional Details : Weight : 0.58900kg
 View more versions of this product
Product Code. 17194211
  • £246.00 / Pack of 3
Estimated Shipment: 07-03-2025
to see stock.
Description
Description
  • Users have the convenient choice between high void-volume depths medium or high-area pleated depth filter
  • Modules can come in different filter specifications to meet the specific needs of an early phase process development
Specifications
Specifications
Spun Polypropylene
Non-sterile
3/Pk.
0.5 μm
46 cm2
9/16 in. Hose Barb