Learn More
Huber™ Minichiller
Minichillers are a space-saving, low-cost and environmentally friendly cooling solution for numerous laboratory applications. By saving cooling water, Minichillers pay for themselves after only a short time.
POA - £3080.00
Products 7
Product Code Brand Model Refrigerant Price Quantity & Availability  
Product Code Brand Model Refrigerant Price Quantity & Availability  
15648298
View Documents
Huber™
3006.0091.98
Minichiller 300-H OLE Natural Refrigerant
POA
Each
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
15713047
View Documents
Huber™
3006.0089.98
Minichiller 300 OLE Natural Refrigerant
POA
Each
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
15723047
View Documents
Huber™
3006.0090.98
Minichiller 300w OLE Natural Refrigerant
POA
Each
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
15733047
View Documents
Huber™
3006.0092.98
Minichiller 300w-H OLE Natural Refrigerant
POA
Each
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
15743047
View Documents
Huber™
3066.0002.98
Minichiller 600 OLE Compact
POA
Each
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
15753047
View Documents
Huber™
3066.0003.98
Minichiller 600-H OLE Compact
POA
Each
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
15773047
View Documents
Huber™
3065.0001.98
£3080.00
Each
Estimated Shipment: 20-05-2025
to see stock.
Add to basket
Description
Description
The compact Minichillers are an economic and environmentally friendly cooling solution for many laboratory applications. A calculation of water and waste water costs shows that a Minichiller can save up to 48,000 L of water in a working week and due to its low procurement price, return on investment is achieved within a very short time.
  • The units have small footprints, only requiring 225 x 360 mm on the laboratory bench and are therefore suitable for cooling reactor blocks, vacuum pumps, rotary evaporators or heat exchangers.
  • Minichillers allow stable application conditions due to constant values for temperature, pressure and throughflow quantities.
  • They are also perfect for continuous operation at ambient temperatures up to + 40°C.
  • Despite the economic purchase price the coolers are well equipped and have a large temperature indicator, fill level indicator, as well as status LEDs for the pump, cooling and heating.
  • A 1 kW heater is optionally available with which the temperature range can be extended to +100°C.
  • Minichillers are available as both air and water cooled models and use natural refrigerants.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Huber™ Minichiller