missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One MiniCollect™ Tube. Lavender Cap, K3E K3EDTA, Non-Sterile
Inner wall is coated with anticoagulant that binds calcium ions and interrupts clotting.
£102.21 - £408.87
Specifications
Additive | K3 E K3 EDTA |
---|---|
Sterility | Non-sterile |
Closure Included | Yes |
Closure Color | Lavender |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15748169
|
Greiner Bio-One
450530 |
0.25, 0.5 mL |
£102.21
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15830146
|
Greiner Bio-One
450530 |
0.25, 0.5 mL |
£408.87
Pack of 2000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15758169
|
Greiner Bio-One
450531 |
1 mL |
£102.21
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Inner wall is coated with spray-dried K3E K3EDTA as an anticoagulant to bind calcium ions and interrupt the clotting cascade.
- Used for hematology.
- Available in different quantities.
Specifications
K3 E K3 EDTA | |
Non-sterile | |
Yes | |
Lavender |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- X500 MiniCollect TUBE 0.25/0.5 ml K3E K3EDTA lave nder cap
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Ethylenediaminetetraacetic Acid Tripotassium Dihydrate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Greiner Bio-One MiniCollect™ Tube. Lavender Cap, K3E K3EDTA, Non-Sterile