missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Water Testing Minikit
Lovibond™ Water Testing Minikit is easy to use and provides highly accurate results.
£51.25 - £74.10
Product Code | Brand | Analytes | Range | Includes | Test Method | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Analytes | Range | Includes | Test Method | Price | Quantity & Availability | |||
10268420
|
Lovibond™
414180 |
Chloride | 5 to 5000mg/L | Flip top case containing a shaker tube, rapid dissolving tablet reagents and instructions. | Tablet count |
£74.10
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10248520
|
Lovibond™
414270 |
Nitrite | 70 to 1500mg/L | Flip top case containing a shaker tube, rapid dissolving tablet reagents and instructions. | Tablet count |
£51.25
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10511931
|
Lovibond™
414340 |
Sulphite (low range) | 2 to 50mg/L | Flip top case containing a shaker tube, rapid dissolving tablet reagents and instructions. | Tablet count |
£51.25
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10024230
|
Lovibond™
414350 |
Sulphite (high range) | 0 to 500mg/L | Flip top case containing a shaker tube, rapid dissolving tablet reagents and instructions. | Tablet count |
£51.25
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10616151
|
Lovibond™
414220 |
Cyanuric Acid | 20 to 200mg/L | Flip top case with calibrated two piece tube, tablet reagents and instructions | Turbidity |
£65.80
Each |
Estimated Shipment: 03-03-2025
Log in to see stock. |
|||
10501931
|
Lovibond™
414320 |
Sulphate (low range) Na2SO4 | 20-200mg/L | Flip top case containing a shaker tube, rapid dissolving tablet reagents and instructions. | Tablet count |
£70.20
Each |
Estimated Shipment: 03-03-2025
Log in to see stock. |
|||
13151701
|
Lovibond™
414100 |
Cleaning Acid Strength | 0.75-10% acid | Flip top case containing a shaker tube, rapid dissolving tablet reagents and instructions. | Tablet count |
£51.25
Each |
Estimated Shipment: 03-03-2025
Log in to see stock. |
|||
10014480
|
Lovibond™
414130 |
Alkalinity Total M | 10 to 500mg/L | Flip top case containing a shaker tube, rapid dissolving tablet reagents and instructions. | Tablet count |
£51.25
Each |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||
10228330
|
Lovibond™
414140 |
Alkalinity-P, CaC03 | 20 to 500mg/L | Flip top case containing a shaker tube, rapid dissolving tablet reagents and instructions. | Tablet count |
£51.25
Each |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||
Description
Lovibond™ Water Testing Minikit is easy to use and provides highly accurate results.
- Easy operation and exact reagent dosing
- High accuracy
- Foil-wrapped Lovibond™ tablet reagents with a minimum guaranteed shelf life of 5 years
- Unrestricted shipment
- Safe storage
Specifications
Approximately 40 Tests | |
Minikit water testing kit |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- MINIKIT TEST CHLORIDE SALINITY AF418
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360-May damage fertility or the unborn child.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : boric acid, potassium chromate, silver nitrate, potassium dichromate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lovibond™ Water Testing Minikit