Learn More
Lovibond™ Water Testing Minikit
Lovibond™ Water Testing Minikit is easy to use and provides highly accurate results.
Brand: Lovibond™ 414130
This item is not returnable.
View return policy
Description
Lovibond™ Water Testing Minikit is easy to use and provides highly accurate results.
- Easy operation and exact reagent dosing
- High accuracy
- Foil-wrapped Lovibond™ tablet reagents with a minimum guaranteed shelf life of 5 years
- Unrestricted shipment
- Safe storage
Specifications
Alkalinity Total M | |
1 Pack | |
Approximately 40 Tests | |
Minikit water testing kit |
Flip top case containing a shaker tube, rapid dissolving tablet reagents and instructions. | |
10 to 500mg/L | |
Tablet count |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- MINIKIT TEST ALKALINITY TOTAL M AF413 .
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : sodium bisulphate
Product Content Correction
Product Title
Lovibond™ Water Testing Minikit > Analytes: Alkalinity Total M
Spot an opportunity for improvement?Share a Content Correction