missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MIP-3 alpha/CCL20 Cell Lysate Human ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EHCCL20CLX10

Additional Details : Weight : 2.50000kg

 View more versions of this product

Product Code. 17894180

  • £4280.00 / Pack of 10
Estimated Shipment: 16-08-2024
to see stock.
Add to basket

Description

Description

The Human MIP-3 alpha/CCL20 ELISA quantitates Hu MIP-3α in human cell or tissue lysates. The assay will exclusively recognize both natural and recombinant Hu MIP-3α. Principle of the method The Human MIP-3α solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Macrophage Inflammatory Protein-3 alpha (MIP-3 alpha), also called CCL20, is expressed in the liver, lungs, lymph nodes and peripheral blood lymphocytes. MIP-3 alpha is strongly up regulated by inflammatory signals, and down regulated by the anti-inflammatory cytokine IL-10. MIP3alpha signals through the G protein-coupled receptor, CCR6, and acts as a chemoattractant to lymphocytes and dendritic cells.
Specifications

Specifications

P78556
1.5 pg/mL
Solid-phase sandwich ELISA
ELISA Kit
Cell Lysates
10 x 96 assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
6364
<12%
Standard, SAV-HRP, Biotinylated Detection Antibody, Adhesive Plate Covers, Wash Buffer, Stop Solution, Pre-coated 96 well plate, Chromogen, Assay Diluent concentrate, Cell Lysis Buffer
10 x 96 Tests
Inflammation
2°C to 8°C
Human
4 hr. 45 min.
0.8 to 600 pg/mL
1.5 pg/mL to 600 pg/mL
Biotin
Chemokine & Receptors
The antibody pair provided in this kit recognizes human MIP-3 alpha.
Human
Colorimetric Microplate Reader
CKb4,Exodus,LARC,MIP-3-alpha,MIP-3 A,MIP3A,SCYA20,ST38
<10%
HRP
RUO
Cell Lysate,20 μL
CKb4, Exodus, LARC, MIP-3-alpha, MIP-3 A, MIP3A, SCYA20, ST38
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers