Learn More
Invitrogen™ MIP-3 alpha/CCL20 Cell Lysate Human ELISA Kit
Sandwich ELISA Kit
£544.00 - £4280.00
Specifications
Accession Number | P78556 |
---|---|
Assay Sensitivity | 1.5 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15387118
|
Invitrogen™
EHCCL20CL |
96 Tests |
£544.00
1 set |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
17894180
|
Invitrogen™
EHCCL20CLX10 |
10 x 96 Tests |
£4280.00
Pack of 10 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
17165379
|
Invitrogen™
EHCCL20CLX5 |
5 x 96 Tests |
£2310.00
Each |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
Description
The Human MIP-3 alpha/CCL20 ELISA quantitates Hu MIP-3α in human cell or tissue lysates. The assay will exclusively recognize both natural and recombinant Hu MIP-3α. Principle of the method The Human MIP-3α solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Macrophage Inflammatory Protein-3 alpha (MIP-3 alpha), also called CCL20, is expressed in the liver, lungs, lymph nodes and peripheral blood lymphocytes. MIP-3 alpha is strongly up regulated by inflammatory signals, and down regulated by the anti-inflammatory cytokine IL-10. MIP3alpha signals through the G protein-coupled receptor, CCR6, and acts as a chemoattractant to lymphocytes and dendritic cells.Specifications
P78556 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Cell Lysates | |
Human | |
Colorimetric Microplate Reader | |
<12% | |
HRP | |
Inflammation | |
Human | |
4 hr. 45 min. |
1.5 pg/mL | |
Biotin | |
Chemokine & Receptors | |
The antibody pair provided in this kit recognizes human MIP-3 alpha. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
6364 | |
<10% | |
RUO | |
2°C to 8°C | |
1 hr. 20 min. |