missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Mixed Cellulose Ester Membranes Sterile Filter

Fisherbrand™ Mixed Celluloses Esters Membranes Sterile Filters are composed of cellulose acetate and cellulose nitrate.

Brand:  Fisherbrand™

68.45 GBP valid until 2024-12-31
Use promo code "21931" to get your promotional price.



Code : A0

Additional Details : Weight : 0.07020kg


 View more versions of this product

Product Code. 11765285

  • £83.30 / Pack of 100
Estimated Shipment: 21-08-2024
to see stock.
Add to basket

Description

Description

  • Widely used membranes in analytical and research applications
  • Naturally hydrophilic, biologically inert and offer thermal stability
  • Characterized by smoother and more uniform surface than pure nitrocellulose membrane which facilitates particles detection, and high retention level
  • Gridded membranes are used in many microbiological techniques include colony counting after incubation
Specifications

Specifications

3.62bar
Microfilter
0.2 μm
19mL/min./cm2
2L/min.
Hydrophilic
Gridded Disc
47 mm
White
MCE
Sterile
Microdialysis of DNA and Proteins, clarification of aqueous solutions during microbiological analysis, identification of Nucleic Acids
Black
100 Pack
Yes
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MBNE MCE WHIT/N 47MM .22µm

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 1

Hazard Statement
  • H228-Flammable solid.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P370+P378-In case of fire: Use powder to extinguish.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose

SDS
Documents

Documents

Certificates
Special Offers

Special Offers