Learn More
Tocris Bioscience™ MK 2206 dihydrochloride
Potent and selective allosteric Akt1, Akt2 and Akt3 inhibitor
Brand:  Tocris Bioscience™ 7850/10
Additional Details : CAS Number : 1032350-13-2 Weight : 0.00970kg
 View more versions of this product
Product Code. 30000034
  • £105.83 / 10mg
Estimated Shipment: 07-08-2024
to see stock.
Description
Description
MK 2206 dihydrochloride is a potent and selective allosteric Akt inhibitor (IC50 values are 5 nM, 12 nM, and 65 nM for Akt1, Akt2, and Akt3, respectively). MK 2206 requires the Pleckstrin homology domain for its activity, and exhibits no inhibitory activity in a panel of 250 tested protein kinases. MK 2206 induces growth inhibition of different cancer cell lines (IC50 in the range 3.4 and 28.6 μmol/L) and enhances antitumor efficacy of a range of standard chemotherapeutics. It synergistically inhibits cell proliferation of human lung and breast cancer cells in combination with Erlotinib (Cat. No. 7194) by suppressing both the Ras/Erk and PI 3-K pathways in vitro and in vivo. MK 2206 can be used to block the IL-22 - Akt signaling pathway for studying paneth cell differentiation.
Specifications
Specifications
8-[4-(1-Aminocyclobutyl)phenyl]-9-phenyl-1,2,4-triazolo[3,4-f][1,6]naphthyridin-3(2H)-one dihydrochloride
10 mg
C25H21N5O.2HCl
1032350-13-2
Akt (Protein Kinase B) Inhibitors
0.98