missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MK 2206 dihydrochloride
Potent and selective allosteric Akt1, Akt2 and Akt3 inhibitor
£105.83 - £396.32
Specifications
Chemical Name or Material | 8-[4-(1-Aminocyclobutyl)phenyl]-9-phenyl-1,2,4-triazolo[3,4-f][1,6]naphthyridin-3(2H)-one dihydrochloride |
---|---|
CAS | 1032350-13-2 |
Target | Akt (Protein Kinase B) Inhibitors |
Molecular Formula | C25H21N5O.2HCl |
Purity | 0.98 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30000034
|
Tocris Bioscience™
7850/10 |
10 mg |
£105.83
10mg |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30000037
|
Tocris Bioscience™
7850/50 |
50 mg |
£396.32
50mg |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MK 2206 dihydrochloride is a potent and selective allosteric Akt inhibitor (IC50 values are 5 nM, 12 nM, and 65 nM for Akt1, Akt2, and Akt3, respectively). MK 2206 requires the Pleckstrin homology domain for its activity, and exhibits no inhibitory activity in a panel of 250 tested protein kinases. MK 2206 induces growth inhibition of different cancer cell lines (IC50 in the range 3.4 and 28.6 μmol/L) and enhances antitumor efficacy of a range of standard chemotherapeutics. It synergistically inhibits cell proliferation of human lung and breast cancer cells in combination with Erlotinib (Cat. No. 7194) by suppressing both the Ras/Erk and PI 3-K pathways in vitro and in vivo. MK 2206 can be used to block the IL-22 - Akt signaling pathway for studying paneth cell differentiation.Specifications
8-[4-(1-Aminocyclobutyl)phenyl]-9-phenyl-1,2,4-triazolo[3,4-f][1,6]naphthyridin-3(2H)-one dihydrochloride | |
Akt (Protein Kinase B) Inhibitors | |
0.98 |
1032350-13-2 | |
C25H21N5O.2HCl |