Learn More
ML-18, MedChemExpress
MedChemExpress ML-18 is a non-peptide bombesin receptor subtype-3 (BRS-3) antagonist with an IC50 of 4.8 μM.
Brand: Medchem Express HY-101844
This item is not returnable.
View return policy
Description
ML-18 is a non-peptide bombesin receptor subtype-3 (BRS-3) antagonist with an IC50 of 4.8 μM.Specifications
ML-18 | |
Light Yellow | |
C32H35N5O5 | |
DMSO : ≥ 100 mg/mL (175.55 mM) | |
569.65 | |
98.44% | |
Solid |
1422269-30-4 | |
Research | |
5 mg | |
O=C([C@@H](NC(NC1=CC=C([N+]([O-])=O)C=C1)=O)CC2=CNC3=CC=CC=C23)NCC4(C5=CC=C(OC)C=C5)CCCCC4 | |
569.65 | |
Research | |
Cancer-Kinase/protease |