missing translation for 'onlineSavingsMsg'
Learn More

ML-18, MedChemExpress

MedChemExpress ML-18 is a non-peptide bombesin receptor subtype-3 (BRS-3) antagonist with an IC50 of 4.8 μM.

£226.27 - £1510.08

Specifications

Chemical Name or Material ML-18
CAS 1422269-30-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C32H35N5O5
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16499996
Encompass_Preferred
View Documents
Medchem Express
HY-101844
50 mg
£1195.48
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16474696
Encompass_Preferred
View Documents
Medchem Express
HY-101844
100 mg
£1510.08
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16453608
Encompass_Preferred
View Documents
Medchem Express
HY-101844
10 mg
£314.60
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16452786
Encompass_Preferred
View Documents
Medchem Express
HY-101844
25 mg
£692.12
25mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16426337
Encompass_Preferred
View Documents
Medchem Express
HY-101844
5 mg
£226.27
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ML-18 is a non-peptide bombesin receptor subtype-3 (BRS-3) antagonist with an IC50 of 4.8 μM.
Specifications

Specifications

ML-18
Light Yellow
C32H35N5O5
O=C([C@@H](NC(NC1=CC=C([N+]([O-])=O)C=C1)=O)CC2=CNC3=CC=CC=C23)NCC4(C5=CC=C(OC)C=C5)CCCCC4
569.65
Research
Cancer-Kinase/protease
1422269-30-4
Research
DMSO : ≥ 100 mg/mL (175.55 mM)
569.65
98.44%
Solid
Videos
Safety and Handling

Safety and Handling

H302∣H315∣H319∣H335

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.