Learn More
Tocris Bioscience™ ML 337
Selective negative allosteric modulator of mGlu3
Brand:  Tocris Bioscience™ 4982/50
393.08 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18708358
  • £492.00 / 50mg
Explore more special offers
Description
Description
ML 337 is a selective negative allosteric modulator of mGlu3 (IC50 = 593 nM). Displays no activity at mGlu1, mGlu2 or mGlu4-8 at concentrations up to 30 μM. Brain penetrant.
Specifications
Specifications
>98%
ML 337
ML 337
50 mg
353.39