missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ML 337
Selective negative allosteric modulator of mGlu3
£130.00 - £492.00
Specifications
Purity | >98% |
---|---|
Product Type | ML 337 |
Molecular Weight (g/mol) | 353.39 |
Inhibitors | ML 337 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18758068
|
Tocris Bioscience™
4982/10 |
10 mg |
£130.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18708358
|
Tocris Bioscience™
4982/50 |
50 mg |
£492.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Selective negative allosteric modulator of mGlu3 (IC50 = 593 nM). Displays no activity at mGlu1, mGlu2 or mGlu4-8 at concentrations up to 30 μM. Brain penetrant.Specifications
>98% | |
ML 337 | |
353.39 | |
ML 337 |
For Research Use Only