Learn More
MMAD-d8, MedChemExpress
MedChemExpress D8-MMAD is a deuterated form of MMAD, which is a microtubule disrupting agent.
Brand:  Medchem Express HY-15581S
Encompass_Preferred
 View more versions of this product
Product Code. 16401539
  • £1510.08 / 1mg
Explore more special offers
Description
Description
D8-MMAD is a deuterated form of MMAD, which is a microtubule disrupting agent.
Specifications
Specifications
MMAD-d8
Off-White
C41H58D8N6O6S
DMSO : ≥ 100 mg/mL (128.35 mM)
779.11
99.12%
Cancer-programmed cell death
1 mg
Research
Demethyldolastatin 10-d8 Monomethylauristatin D-d8 Monomethyl Dolastatin 10-d8
O=C(N[C@H](C1=NC=CS1)CC2=CC=CC=C2)[C@H](C)[C@@H](OC)[C@H]3N(C(C[C@@H](OC)[C@H]([C@@H](C)CC)N(C([C@@](C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])([2H])NC([C@H](C(C)C)NC)=O)=O)C)=O)CCC3
779.11
Research
Solid