missing translation for 'onlineSavingsMsg'
Learn More

MMAD-d8, MedChemExpress

MedChemExpress D8-MMAD is a deuterated form of MMAD, which is a microtubule disrupting agent.

£1510.08 - £6454.14

Specifications

Chemical Name or Material MMAD-d8
Color Off-White
Purity Grade Notes Research
Molecular Formula C41H58D8N6O6S
Synonym Demethyldolastatin 10-d8 Monomethylauristatin D-d8 Monomethyl Dolastatin 10-d8
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16489207
Encompass_Preferred
View Documents
Medchem Express
HY-15581S
5 mg
£3432.77
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16466648
Encompass_Preferred
View Documents
Medchem Express
HY-15581S
10 mg
£6454.14
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16401539
Encompass_Preferred
View Documents
Medchem Express
HY-15581S
1 mg
£1510.08
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

D8-MMAD is a deuterated form of MMAD, which is a microtubule disrupting agent.
Specifications

Specifications

MMAD-d8
Research
Demethyldolastatin 10-d8 Monomethylauristatin D-d8 Monomethyl Dolastatin 10-d8
O=C(N[C@H](C1=NC=CS1)CC2=CC=CC=C2)[C@H](C)[C@@H](OC)[C@H]3N(C(C[C@@H](OC)[C@H]([C@@H](C)CC)N(C([C@@](C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])([2H])NC([C@H](C(C)C)NC)=O)=O)C)=O)CCC3
779.11
Research
Solid
Off-White
C41H58D8N6O6S
DMSO : ≥ 100 mg/mL (128.35 mM)
779.11
99.12%
Cancer-programmed cell death
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣The compound is unstable in solutions, freshly prepared is recommended.

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣The compound is unstable in solutions, freshly prepared is recommended.

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.