Learn More
Invitrogen™ mMESSAGE mMACHINE™ T3 Transcription Kit
Obtain 10–50 times yield of conventional in vitro transcription reactions
£540.00
Product Code | Brand | Product Type | No. of Reactions | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | No. of Reactions | Price | Quantity & Availability | ||||
10665605
|
Invitrogen™
AM1348 |
T3 In Vitro Transcription kit | 25 Reactions |
£540.00
25 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Ambion mMESSAGE mMACHINE High Yield Capped RNA Transcription Kits use Ambion's patented high-yield transcription technology for the routine synthesis of 15 - 35μg of 7-methyl guanosine capped RNA from 1μg of template DNA. Each kit includes sufficient reagents for 25 reactions.
- High yields of capped RNA—up to 39μg of RNA per reaction
- Extra convenience—cap analog-nucleotide mix provided
- Simplified reaction format
- Synthesized transcripts ideal for microinjection, in vitro translation, or transfection in a 20μL reaction during a 2 hour incubation, mMESSAGE mMACHINE High Yield Capped RNA Transcription will yield up to 35μg of capped RNA
- ™
- 10–50 times yield obtained with conventional in vitro transcription reactions
- Ratio of cap analog to GTP has been optimized to allow compromise between yield (15 - 35μg) and proportion of transcripts that are capped (80%)
- Contain a LiCl precipitation solution, efficient for separating proteins and unincorporated nucleotides (including free cap analog) from the capped RNA, allowing an increased efficiency of translation
- Optimized for use with polymerases included in kit
- Contains 57U of cap analog
For Research Use Only. Not for use in diagnostic procedures.