man
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Clamps and Supports
Clamps
Bochem™ Mohr Pinch Cocks
10459582
Print
Bochem™ Mohr Pinch Cocks
Length: 40mm
Brand:
Bochem™ 12519
View more versions of this product
Product Code.
10459582
Explore more special offers
This item is not returnable.
View return policy
Description
Specifications
Product Suggestions
Videos
SDS
Documents
Certificates
Special Offers
Description
Description
Specifications
Specifications
Quantity
1 Ea.
Material
Brass
Length (Metric)
40 mm
Type
Pinchcock
Diameter (Metric)
15 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers