missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ Mohr Pinch Cocks
£2.85 - £5.45
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10459582
|
Bochem™
12519 |
40 mm |
£3.85
Each |
Estimated Shipment: 25-10-2024
Log in to see stock. |
|||||
10088760
|
Bochem™
12520 |
50 mm |
£2.85
Each |
Estimated Shipment: 23-10-2024
Log in to see stock. |
|||||
10180711
|
Bochem™
12522 |
70 mm |
£5.45
Each |
Estimated Shipment: 17-10-2024
Log in to see stock. |
|||||
Specifications
1 Ea. | |
Pinchcock | |
Brass |