Learn More
Invitrogen™ ATTO-TAG™ FQ Derivatization Reagent (FQ; 3-(2-Furoyl)quinoline-2-Carboxaldehyde)
Allows ultrasensitive detection of primary amines
Brand: Invitrogen™ A10192
Code : 50
Additional Details : Weight : 0.00052kg
Description
Recently, researchers described a simple method in which the ATTO-TAG FQ reagent was used to derivatize very dilute solutions (10-8 M) of peptides. Available as standalone product or as component of an amine-derivatization kit (A-2334). Particularly well-suited to derivatization of peptides, amino acids, aminated sugars and other amine-containing molecules for detection by capillary electrophoresis with laser-induced fluorescence, as its nonfluorescent in unreacted state and reacts specifically with amines to form highly fluorescent conjugates.
Anion Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature
Specifications
Store in freezer (-5 to -30°C) and protect from light. | |
Aldehyde | |
Solid | |
10 mg | |
Primary Amine | |
ATTO-TAG FQ |
Amine | |
Room Temperature | |
ATTO-TAG™ | |
Detection of primary amines | |
Derivatization Kits and Reagents |
For Research Use Only. Not for use in diagnostic procedures.