missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BCECF, AM (2',7'-Bis-(2-Carboxyethyl)-5-(and-6)-Carboxyfluorescein, Acetoxymethyl Ester)
For measuring changes in the cytosolic pH of most cells
Brand: Invitrogen™ B1150
147.84 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
- With pKa of approximately 6.98, cell-permeant, dual-excitation ratiometric pH indicator, AM is ideal for measuring changes in cytosolic pH of most cells
- Intracellular pH measurements with BCECF are made by determining pH-dependent ratio of emission intensity (detected at 535nm) when dye is excited at approximately 490nm versus emission intensity when excited at its isobestic point of approximately 440nm
Cell Adhesion, Cell Analysis, Cell Metabolism, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Cell-Based Ion Channel Assays, Drug Discovery and Development, Ion Channel Biology, Ionic Homeostasis and Signaling, Target and Lead Identification and Validation, pH Detection
Order Info
Shipping Condition: Room temperature
Specifications
Store in freezer -5°C to -30°C. | |
Microplate Reader | |
pH Indicator | |
Room Temperature |
Fluorescence | |
BCECF AM pH Indicator | |
1mg |
For Research Use Only. Not for use in diagnostic procedures.