Learn More
Invitrogen™ FocalCheck™ Fluorescence Microscope Test Slide #1, for alignment, intensity, & calibration
For confocal and fluorescence microscope calibration
Brand: Invitrogen™ F36909
Code : 50
Additional Details : Weight : 0.02000kg
Description
The FocalCheck fluorescence microscope test slide #1 is specifically designed for calibrating fluorescence microscope systems and evaluating system and filter performance. The slide contains 10 sample areas coated with FocalCheck ring stained microspheres ranging in size from 0.5-15μm that provide a visual representation of instrument misalignment or other aberrations in row A; row B contains a series of microspheres with fluorescence intensities ranging from 0.667-100%.
Cell Analysis, Cell Cycle, Cell Viability, Proliferation and Function, Cellular Imaging, Immunofluorescence (IF)
Order Info
Shipping Condition: Room temperature
Specifications
Fluorescence Microscope Test Slide | |
Confocal Microscope Calibration, Fluorescence Microscope Calibration | |
Suspension Beads | |
1 Ea. |
FocalCheck | |
Assorted Colors | |
Store at room temperature. | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.