Learn More
Invitrogen™ LIVE/DEAD™ Cell Imaging Kit (488/570)
Sensitive two-color fluorescence cell viability assay optimized for FITC and Texas Red™ filters
Brand: Invitrogen™ R37601
228.76 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Includes: Component A: 10 x 1mL vials; Component B: 1 x 10 vials, dried down
Description
- Allows discrimination between live and dead cells with two probes that measure recognized parameters of cytotoxicity and cell viability: intracellular esterase activity and plasma membrane integrity
- Based on cell-permeable dye for staining of live cells and cell-impermeable dye for staining of dead and dying cells
- Adapted for imaging platforms by optimizing for common green and red imaging filters used with FITC and Texas Red
- Live cell component produces intense, uniform green fluorescence in live cells (ex/em 488 nm/515nm
- Dead cell component produces predominantly nuclear red fluorescence (ex/em 570nm/602nm) in cells with compromised cell membranes, strong indicator of cell death and cytotoxicity
- Sensitive detection of key measures of cell health: viability, live/dead ratio, and cytotoxicity
- Ease of use with minimal dilutions
- Live cells are distinguished by presence of ubiquitous intracellular esterase activity as determined by the enzymatic conversion of virtually nonfluorescent cell-permeant calcein AM to intensely fluorescent calcein, which is well-retained within live cells
- Red component is cell-impermeant and therefore only enters cells with damaged membranes
- In dying and dead cells bright red fluorescence is generated upon binding to DNA
- Background fluorescence levels are inherently low with this assay technique because dyes are virtually nonfluorescent before interacting with cells
Cell Analysis, Cell Counting and Viability, Cell Counting, Viability, and Cryopreservation, Cell Culture, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function
Order Info
Shipping Condition: Dry Ice

Specifications
Mammalian Cells | |
• Component A: 10 x 1 mL vials • Component B: 1 x 10 vials (dried down) Store at -20°C |
|
Fluorescence | |
Floid Cell Imaging System, Fluorescence Microscope | |
Other Label(s) or Dye(s) | |
488, 570 nm | |
Tube(s) | |
LIVE/DEAD™, Molecular Probes™ | |
Dry Ice |
Red, Green | |
LIVE/DEAD™ Cell Imaging Kit (488/570) | |
Viability Assay | |
Cell Imaging Kit | |
515, 602 nm | |
Frozen | |
15 min | |
1 mL kit |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3079927 | Certificate of Analysis | 29/12/2024 | R37601_LITL18 PROMO,R37601 |
3079918 | Certificate of Analysis | 22/12/2024 | R37601_LITL18 PROMO,R37601 |
2994103 | Certificate of Analysis | 25/09/2024 | R37601_LITL18 PROMO,R37601 |
2983235 | Certificate of Analysis | 22/08/2024 | R37601_LITL18 PROMO,R37601 |
2971024 | Certificate of Analysis | 11/07/2024 | R37601_LITL18 PROMO,R37601 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.