missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LIVE BacLight™ Bacterial Gram Stain Kit, for microscopy & quantitative assays

One step assay

Brand:  Invitrogen™ L7005

435.40 GBP valid until 2025-06-30
Use promo code "24074" to get your promotional price.


Product Code. 10217302

  • £622.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

The LIVE BacLight™ Bacterial Gram Stain Kit provides a novel one-step fluorescence assay for the determination of gram sign in living bacteria. When gram negative and gram-positive bacterial are simultaneously stained with the membrane-permeant SYTO 9 dye and hexidium iodide, gram-negative bacteria fluoresce green and gram-positive bacteria fluoresce red.

Any physiological buffer between pH 7.0–8.0, including PBS, can be used to dilute the SYTO dyes for the staining solution.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Green, Red
LIVE BacLight™ Bacterial Gram Stain Kit, for microscopy and quantitative assays
Viability Assay
Bacterial Stain Kit
480-700 nm
Tube(s), 96-well plate, Slide(s)
1 kit
Store in freezer at -5°C to -30°C and protect from light.
Fluorescence
Fluorescence Microscope, Fluorometer, Flow Cytometer, Microplate Reader
Other Label(s) or Dye(s)
470 nm
BacLight™
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2906446 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog NumberL7005
Lot Number2674024 Certificate TypeCertificate of Analysis Date14/08/2023 Catalog NumberL7005
Lot Number2525766 Certificate TypeCertificate of Analysis Date28/09/2022 Catalog NumberL7005
Lot Number2441337 Certificate TypeCertificate of Analysis Date17/01/2022 Catalog NumberL7005
Lot Number2395116 Certificate TypeCertificate of Analysis Date24/08/2021 Catalog NumberL7005
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ LIVE BacLight™ Bacterial Gram Stain Kit, for microscopy & quantitative assays > 1 kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.