Learn More
Invitrogen™ Measure-IT™ Thiol Assay Kit
Provides easy and accurate quantitation of thiol
Brand: Invitrogen™ M30550
Code : 50
Additional Details : Weight : 0.00525kg Transport : UN number : 3316 Chem class : 9 Pack group : III
Description
- Linear range of 0.05-5μM thiol, making it up to 400 times more sensitive than colorimetric methods based on Ellman's reagent
- Assay is performed at room temperature; maximum fluorescence signal is attained in 5 minutes and is stable for at least 1 hour
Biochemical Toxicology Assays, Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Drug Discovery and Development, Target-Based ADME/Tox Assays
Order Info
Shipping Condition: Room Temperature
Specifications
Thiol Assay Kit |
For Research Use Only. Not for use in diagnostic procedures.