Learn More
Invitrogen™ Naphthalene-2,3-Dicarboxaldehyde (NDA)
Tagets amines
Brand: Invitrogen™ N1138
Code : 50
Additional Details : Weight : 0.00450kg
Description
- Aromatic dialdehyde naphthalene-2,3-dicarboxaldehyde (NDA) is essentially nonfluorescent until reacted with primary amine in presence of excess cyanide or thiol, to yield fluorescent isoindole with excitation/emission maxima approximately 419/493 nm
Anion Detection, Cell Analysis, Cell Viability, Proliferation and Function, Glutathione Detection, Ionic Homeostasis and Signaling, Nitro-Oxidative Stress, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature
Specifications
Store at room temperature and protect from light. | |
Room Temperature | |
Primary Amine | |
NDA (naphthalene-2,3-dicarboxaldehyde) |
Fluorescent | |
Solid | |
Naphthalene-2,3-Dicarboxaldehyde (NDA) |
For Research Use Only. Not for use in diagnostic procedures.