Learn More
Invitrogen™ Zymosan A S. cerevisiae BioParticles™, fluorescein conjugate
Study phagocytosis by fluorescence microscopy, quantitative spectrofluorometry, and flow cytometry
Brand: Invitrogen™ Z2841
202.16 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
- Fluorescence of fluorescein-labeled BioParticles conjugates, partially quenched in acidic environments, fluorescence of the Alexa Fluor, BODIPY™ FL, tetramethylrhodamine and Texas Red™ dye conjugates is uniformly intense over the pH range from 3 to 10
- Label (Ex/Em): Fluorescein (approx. 494/518nm)
- Particle: Zymosan (S. cerevisiae)
- Provided as lyophilized powders
- Approximately 3 x 108 E. coli or S. aureus particles per mg solid and approximately 2 x 107 zymosan particles per mg solid
- Conjugates can be reconstituted in the buffer of choice for use in phagocytosis assays
- Fluorescence of BioParticles conjugates that are bound to surface of cell (but not internalized) can be quenched by ethidium bromide, trypan blue, or other quenchers
- Addition to cellular applications, fluorescent BioParticles conjugates may be effective as flow cytometry calibration references when sorting bacteria and yeast mutants
- Small particles may also be useful references for light scattering studies because their sizes and shapes differ in characteristic ways
Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Immunofluorescence (IF), Immunofluorescence Staining and Detection
Order Info
Shipping Condition: Room Temperature

Specifications
Store in freezer (-5 to -30°C) and protect from light. | |
Fluorescence Microscope | |
FITC (Fluorescein) | |
3 to 10 | |
10 mg | |
S. cerevisiae |
Fluorescence | |
Bioparticle Conjugate | |
Solid | |
BioParticles™ | |
Room Temperature |
Suggestions de produits
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
For Research Use Only. Not for use in diagnostic procedures.