missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieves, 3 Å, 4 to 8mesh, Honeywell Fluka™

Beads, 4-8 mesh

£139.36 - £327.86

Chemical Identifiers

CAS 308080-99-1
Molecular Formula AlNaO6Si2
Molecular Weight (g/mol) 202.135
MDL Number MFCD00131613
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
PubChem CID 19758701
IUPAC Name aluminum;sodium;dioxido(oxo)silane
SMILES [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15661840
View Documents
Honeywell-Fluka
208574-1KG
1 kg
£139.36
1kg
Please to purchase this item. Need a web account? Register with us today!
15671840
View Documents
Honeywell-Fluka
208574-5KG
5 kg
£327.86
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

308080-99-1
202.135
URGAHOPLAPQHLN-UHFFFAOYSA-N
aluminum;sodium;dioxido(oxo)silane
AlNaO6Si2
MFCD00131613
19758701
[O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]

Specifications

4 to 8mesh
AlNaO6Si2
MFCD00131613
URGAHOPLAPQHLN-UHFFFAOYSA-N
aluminum;sodium;dioxido(oxo)silane
19758701
308080-99-1
KnNa12-n[(AlO2)12(SiO2)12] · xH2O
NONH for all modes of transport
[O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
202.135
Molecular Sieves, 3 Å
Videos
SDS
Documents

Documents

Product Certifications