Learn More
Molybdenum silicide, Mo 61% min, typically 99.5% (metals basis), Thermo Scientific Chemicals
Description
In electrical resistors, in protective coatings at high temperatures, and in engine parts in space vehiclesMolybdenum silicide is used in electric furnaces, glass, steel, electronics and ceramics industries. It finds application in heat treatment materials. It is also used in microelectronics as a contact material. Further, it serves as a dry lubricant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- In electrical resistors, in protective coatings at high temperatures, and in engine parts in space vehicles

Specifications
Specifications
Melting Point | 1870°C to 2030°C |
Physical Form | Powder |
Quantity | 250 g |
Solubility Information | Insoluble in acids and aqua regia,however,it is soluble in HF and HNO3Soluble in nitric acid and hydrofluoric acid; Insoluble in water most acids. |
Formula Weight | 152.13 |
Percent Purity | 99.5% (typically) |
Concentration or Composition (by Analyte or Components) | Mo 61% min. |
Odor | Odorless |
Density | 6.31 g/mL |
Assay | (metals basis) |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
U30K004 | Certificate of Analysis | 30/09/2024 | 011549.30,011549.18 |
N21I061 | Certificate of Analysis | 12/04/2022 | 011549.30,011549.18 |
C22X038 | Certificate of Analysis | 17/11/2021 | 011549.30,011549.18 |
M05C005 | Certificate of Analysis | 17/11/2021 | 011549.30,011549.18 |
X23G028 | Certificate of Analysis | 17/11/2021 | 011549.30,011549.18 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.