Learn More
mono-tert-Butyl malonate, 95%, Thermo Scientific Chemicals
£23.60 - £120.00
Chemical Identifiers
CAS | 40052-13-9 |
---|---|
Molecular Formula | C7H12O4 |
Molecular Weight (g/mol) | 160.17 |
InChI Key | NGGGZUAEOKRHMA-UHFFFAOYSA-N |
Synonym | 3-tert-butoxy-3-oxopropanoic acid, malonic acid mono-tert-butyl ester, mono-tert-butyl malonate, 2-tert-butoxycarbonyl acetic acid, tert-butyl hydrogen malonate, mono-t-butyl malonate, pubchem13754, malonicacidmono-tert-butylester, malonic acid mono t-butyl ester |
PubChem CID | 545853 |
IUPAC Name | 3-[(2-methylpropan-2-yl)oxy]-3-oxopropanoic acid |
SMILES | CC(C)(C)OC(=O)CC(=O)O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
40052-13-9 | |
160.17 | |
3-tert-butoxy-3-oxopropanoic acid, malonic acid mono-tert-butyl ester, mono-tert-butyl malonate, 2-tert-butoxycarbonyl acetic acid, tert-butyl hydrogen malonate, mono-t-butyl malonate, pubchem13754, malonicacidmono-tert-butylester, malonic acid mono t-butyl ester | |
3-[(2-methylpropan-2-yl)oxy]-3-oxopropanoic acid |
C7H12O4 | |
NGGGZUAEOKRHMA-UHFFFAOYSA-N | |
545853 | |
CC(C)(C)OC(=O)CC(=O)O |
Specifications
40052-13-9 | |
100.0 | |
Colorless | |
Authentic | |
C7H12O4 | |
NGGGZUAEOKRHMA-UHFFFAOYSA-N | |
3-[(2-methylpropan-2-yl)oxy]-3-oxopropanoic acid | |
545853 | |
95% | |
mono-tert-Butyl malonate |
94.0 | |
19°C to 20°C | |
90°C (2.0 mmHg) | |
95% | |
3-tert-butoxy-3-oxopropanoic acid, malonic acid mono-tert-butyl ester, mono-tert-butyl malonate, 2-tert-butoxycarbonyl acetic acid, tert-butyl hydrogen malonate, mono-t-butyl malonate, pubchem13754, malonicacidmono-tert-butylester, malonic acid mono t-butyl ester | |
CC(C)(C)OC(=O)CC(=O)O | |
160.17 | |
160.17 | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Fatal if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
- mono-tert-Butyl malonate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only