missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Velp Scientifica™ MonoAluBlock™ Block with Forty Positions
For Use With: AREX-6 Hotplate Stirrers
Brand: Velp Scientifica™ A00000341
This item is not returnable.
View return policy
Description
- Used to perform numerous reactions reducing time and bench space
- For use with Velp Series 6 Hotplate Stirrers
Specifications
12 mm | |
14 mm | |
Block |
AREX-6 Hotplate Stirrers | |
40 Positions | |
14 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Velp Scientifica™ MonoAluBlock™ Block with Forty Positions > Holds: 12 x 14 mm Vials
Spot an opportunity for improvement?Share a Content Correction