missing translation for 'onlineSavingsMsg'
Learn More
Montelukast sodium, 98%, Thermo Scientific Chemicals Product Code.: 15555196

Montelukast sodium, 98%, Thermo Scientific Chemicals

Product Code. 15555196
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 15555196

Brand: Thermo Scientific Acros 456440010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 151767-02-1
Quantity 1 g
Molecular Formula C35H35ClNNaO3S
MDL Number MFCD00931431
InChI Key LBFBRXGCXUHRJY-HKHDRNBDSA-M
Melting Point 115.0°C
SMILES [Na+].CC(C)(O)C1=CC=CC=C1CC[C@@H](SCC1(CC([O-])=O)CC1)C1=CC=CC(\C=C\C2=CC=C3C=CC(Cl)=CC3=N2)=C1
IUPAC Name sodium;2-[1-[[(1R)-1-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(2-hydroxypropan-2-yl)phenyl]propyl]sulfanylmethyl]cyclopropyl]acetate
Solubility Soluble in water
Molecular Weight (g/mol) 608.17
PubChem CID 23663996
ChEBI CHEBI:6993
Formula Weight 608.17
Percent Purity 98%
Chemical Name or Material Montelukast sodium
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465026 Certificate TypeCertificate of Analysis Date14/08/2024 Catalog Number456440010
Lot NumberA0441303 Certificate TypeCertificate of Analysis Date02/11/2023 Catalog Number456440010
Lot NumberA0402784 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number456440010
Lot NumberA0398847 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number456440010
Lot NumberA0413287 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number456440010
compliance-icons
Product Identifier
  • Montelukast sodium
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Montelukast sodium, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.