Learn More
Motic™ Moticam A1
The Moticam A1 is ideal for newcomers and advanced enthusiasts who are looking for an affordable digital microscope instrument. Capture everything you need at 720p and work at a steady frame rate with all accessories included.
Brand:  Motic™ 1100600101412
Product Code. 17867960
  • £199.00 / Each
Estimated Shipment: 01-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • C-mount for lens
  • MotiConnect software is available for free download (with free updates included)
  • Live Display Modes: 1280 x 720 px, 1024 x 768 px
Specifications
Specifications
Camera
Focusable Lens, 30 mm and 38 mm Eyepiece Adapters, Calibration Slide, Macro Viewing Tube, C-ring, USB Cable, Dust Cap
1/3.1 in.
Moticam A
sCMOS
30 fps
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Motic™ Moticam A1 > Moticam A1; Frequency (Frame Rate): 30 fps; Sensor Types: sCMOS; Product Line: Moticam A Series; Includes: Focusable Lens, 30 mm and 38 mm Eyepiece Adapters, Calibration Slide, Macro Viewing Tube, C-ring, USB Cable, Dust Cap