Learn More
Fisherbrand™ Motorised Pipette Controller
A powerful, lightweight and ergonomically designed pipetting controller for all commonly used glass or plastic pipettes.
Brand:  Fisherbrand™ FB_Accumaxfiller_EU
Product Code. 15840053
  • £365.00 / Each
Explore more special offers
Description
Description
  • Powerful yet quiet pump
  • Ergonomic design
  • Soft grip push buttons
  • Double safety valve
  • Autoclavable pipette cone
  • two speed modes High, Low and Gravity
  • Switch located on slide part of controller ensuring ease of operation
  • LED indicator lights up in red when battery is low
  • Can be operated continuously for 8 hours
  • Replaceable membrane Filter to protect device from contaminants being drawn into pipette
  • Can be used while it is charging
Pipet, filler, liquid handling
Specifications
Specifications
1000mAh NiMH batteries
Blue
Pipette Controller
Hydrophobic PTFE membrane filter
Glass or plastic serological pipettes
Two (High and Low, along with Gravity drain)
2 x 1.2V / 1000mAh NiMH batteries
Cone shape
Soft Push Button
0.1 to 200 mL
Push button
Pipette filler, Charger (EU plug), Desktop stand, Wall mount bracket, Two spare membrane filters: 0.20μm and 0.45μm, Instruction manual, Batteries
2 to 3 hours
1.2 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Motorised Pipette Controller > Motorised Pipette Controller