Learn More
Gibco™ Mouse Betacellulin Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand:  Gibco™ 315-21-100UG
 View more versions of this product
Product Code. 17832833
  • £482.00 / 100µg
Explore more special offers
Description
Description
The protein encoded by this gene is a member of the EGF family of growth factors. It is synthesized primarily as a transmembrane precursor, which is then processed to mature molecule by proteolytic events. This protein is a ligand for the EGF receptor.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Q05928
protein with no preservative
9
100 μg
-20°C
<1 EU/μg
BTC
Unconjugated
DGNTTRTPET NGSLCGAPGE NCTGTTPRQK VKTHFSRCPK QYKHYCIHGR CRFVVDEQTP SCICEKGYFG ARCERVDLFY
≥ 98% by SDS-PAGE gel and HPLC analyses.
Functional Assay
12223
Mouse Betacellulin
E. coli
RUO
Bcn; betacellulin; betacellulin, epidermal growth factor family member; BTC; OTTHUMP00000160600; OTTHUMP00000219057; Probetacellulin
Mouse
Recombinant
Lyophilized