missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse Betacellulin Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£65.00 - £2410.00
Specifications
Accession Number | Q05928 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 12223 |
Molecular Weight (g/mol) | 9 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17832833
|
Gibco™
315-21-100UG |
100 μg |
£482.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17862013
|
Gibco™
315-21-1MG |
1 mg |
£2410.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17812633
|
Gibco™
315-21-20UG |
20 μg |
£158.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17862313
|
Gibco™
315-21-250UG |
250 μg |
£842.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17832633
|
Gibco™
315-21-500UG |
500 μg |
£1442.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17832443
|
Gibco™
315-21-5UG |
5 μg |
£65.00
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a member of the EGF family of growth factors. It is synthesized primarily as a transmembrane precursor, which is then processed to mature molecule by proteolytic events. This protein is a ligand for the EGF receptor.Specifications
Q05928 | |
protein with no preservative | |
9 | |
E. coli | |
RUO | |
Bcn; betacellulin; betacellulin, epidermal growth factor family member; BTC; OTTHUMP00000160600; OTTHUMP00000219057; Probetacellulin | |
Mouse | |
Recombinant | |
Lyophilized |
Functional Assay | |
12223 | |
Mouse Betacellulin | |
-20°C | |
<1 EU/μg | |
BTC | |
Unconjugated | |
DGNTTRTPET NGSLCGAPGE NCTGTTPRQK VKTHFSRCPK QYKHYCIHGR CRFVVDEQTP SCICEKGYFG ARCERVDLFY | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |