missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse Betacellulin Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 315-21-250UG
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the EGF family of growth factors. It is synthesized primarily as a transmembrane precursor, which is then processed to mature molecule by proteolytic events. This protein is a ligand for the EGF receptor.Specifications
Q05928 | |
protein with no preservative | |
9 | |
250 μg | |
-20°C | |
<1 EU/μg | |
BTC | |
Unconjugated | |
DGNTTRTPET NGSLCGAPGE NCTGTTPRQK VKTHFSRCPK QYKHYCIHGR CRFVVDEQTP SCICEKGYFG ARCERVDLFY | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
12223 | |
Mouse Betacellulin | |
E. coli | |
RUO | |
Bcn; betacellulin; betacellulin, epidermal growth factor family member; BTC; OTTHUMP00000160600; OTTHUMP00000219057; Probetacellulin | |
Mouse | |
Recombinant | |
Lyophilized |