missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse EGF Recombinant Protein

Recombinant Mouse Epidermal Growth Factor (EGF) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PMG8045

 View more versions of this product

Product Code. 10543274

  • £75.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Mouse EGF Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

EGF has a profound effect on the differentiation of specific cells in vivo and is a potent mitogenic factor for a variety of cultured cells of both ectodermal and mesodermal origin.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by measuring the dose-dependent proliferation of BALB/c 3T3 cells
  • This product is supplied lyophilized, carrier-free

Shipping Condition: Wet Ice.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

AAS83395, P01133
AAS83395, P01133
Sequential Chromatography
EGF, Pro-epidermal growth factor precursor, epidermal growth factor
53 aa
Untagged
Carrier-Free
25 μg
E. coli
Full Length, Recombinant, Ligand
EGF (Epidermal Growth Factor)
Gibco™
Cell Culture
6 kDa
25 μg
EGF
Mouse
This product is supplied lyophilized, carrier-free.
This lyophilized preparation is stable at 2 to 8°C, but should be kept at –20°C for long term storage, preferably desiccated. Upon reconstitution, the preparation is stable for up to one week at 2 to 8°C. For maximal stability, apportion the reconstituted preparation into working aliquots and store at –20°C. Avoid repeated freeze⁄thaw cycles.
Very Low
Wet Ice
Growth Factors & Receptors
Lyophilized
Stem Cell Research, Oncology
95 %
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.