missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse EGF Recombinant Protein
Recombinant Mouse Epidermal Growth Factor (EGF) is a bioactive protein intended for use in cell culture applications
£62.00 - £522.00
Specifications
Accession Number | AAS83395, P01133 |
---|---|
For Use With (Application) | Cell Culture |
Molecular Weight (g/mol) | 6 kDa |
Purification Method | Sequential Chromatography |
Gene Alias | EGF, Pro-epidermal growth factor precursor, epidermal growth factor |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10134603
|
Gibco™
PMG8044 |
10μg |
£62.00
Each |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10543274
|
Gibco™
PMG8045 |
25μg |
£75.00
Each |
Estimated Shipment: 30-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10564614
|
Gibco™
PMG8041 |
100μg |
£94.25
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10144983
|
Gibco™
PMG8043 |
1mg |
£522.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Mouse EGF Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
EGF has a profound effect on the differentiation of specific cells in vivo and is a potent mitogenic factor for a variety of cultured cells of both ectodermal and mesodermal origin.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by measuring the dose-dependent proliferation of BALB/c 3T3 cells
- This product is supplied lyophilized, carrier-free
Shipping Condition: Wet Ice.
Specifications
AAS83395, P01133 | |
6 kDa | |
EGF, Pro-epidermal growth factor precursor, epidermal growth factor | |
53 aa | |
Untagged | |
Carrier-Free | |
Wet Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Stem Cell Research, Oncology | |
95 % |
Cell Culture | |
Sequential Chromatography | |
EGF | |
Mouse | |
This product is supplied lyophilized, carrier-free. This lyophilized preparation is stable at 2 to 8°C, but should be kept at –20°C for long term storage, preferably desiccated. Upon reconstitution, the preparation is stable for up to one week at 2 to 8°C. For maximal stability, apportion the reconstituted preparation into working aliquots and store at –20°C. Avoid repeated freeze⁄thaw cycles. |
|
Very Low | |
E. coli | |
Full Length, Recombinant, Ligand | |
EGF (Epidermal Growth Factor) | |
Gibco™ |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.