missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse FGF-23 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 450-55-20UG
Additional Details : Weight : 0.16500kg
Description
The protein encoded by this gene is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The product of this gene inhibits renal tubular phosphate transport. This gene was identified by its mutations associated with autosomal dominant hypophosphatemic rickets, an inherited phosphate wasting disorder. Abnormally high level expression of this gene was found in oncogenic hypophosphatemic osteomalacia, a phenotypically similar disease caused by abnormal phosphate metabolism. Mutations in this gene have also been shown to cause familial tumoral calcinosis with hyperphosphatemia.Specifications
Q9EPC2 | |
protein with no preservative | |
25.5 | |
20 μg | |
-20°C | |
<1 EU/μg | |
FGF23 | |
Unconjugated | |
MYPDTSPLLG SNWGSLTHLY TATARTSYHL QIHRDGHVDG TPHQTIYSAL MITSEDAGSV VITGAMTRRF LCMDLHGNIF GSLHFSPENC KFRQWTLENG YDVYLSQKHH YLVSLGRAKR IFQPGTNPPP FSQFLARRNE VPLLHFYTVR PRRHTRSAED PPERDPLNVL KPRPRATPVP VSCSRELPSA EEGGPAASDP LGVLRRGRGD ARGGAGGADR CRPFPRFV | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
64654 | |
Mouse FGF-23 | |
E. coli | |
RUO | |
ADHR; FGF; Fgf23; FGF-23; FGFN; Fibroblast growth factor; Fibroblast growth factor 23; Fibroblast growth factor 23 C-terminal peptide; Fibroblast growth factor 23 N-terminal peptide; HPDR2; HYPF; Phosphatonin; PHPTC; tumor-derived hypophosphatemia inducing factor; Tumor-derived hypophosphatemia-inducing factor; UNQ3027/PRO9828 | |
Mouse | |
Recombinant | |
Lyophilized |