missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Follistatin-related Gene Protein/FLRG Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems MAB1255
This item is not returnable.
View return policy
Description
Follistatin-related Gene Protein/FLRG/Fstl3 Monoclonal specifically detects Follistatin-related Gene Protein/FLRG/Fstl3 in Mouse samples. It is validated for Western Blot.Specifications
Follistatin-related Gene Protein/FLRG/Fstl3 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q9EQC7 | |
FSTL3 | |
Mouse myeloma cell line NS0-derived recombinant mouse FLRG Val24-Val256 Accession # Q9EQC7 | |
500 μg | |
Primary | |
Detects mouse FLRG in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows 50-100% cross-reactivity with recombinant human FLRG. | |
Mouse | |
Purified |
Western Blot | |
210725 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLRGFSRP, Follistatin-like 3, follistatin-like 3 (secreted glycoprotein), Follistatin-like protein 3, Follistatin-related gene protein, follistatin-related protein 3, FSTL3 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10272 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
For Research Use Only