missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Follistatin-related Gene Protein/FLRG Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
£146.86 - £442.75
Specifications
Antigen | Follistatin-related Gene Protein/FLRG/Fstl3 |
---|---|
Clone | 210725 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18713979
|
R&D Systems
MAB1255-SP |
25 μg |
£146.86
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18140541
|
R&D Systems
MAB1255 |
500 μg |
£442.75
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Follistatin-related Gene Protein/FLRG/Fstl3 Monoclonal specifically detects Follistatin-related Gene Protein/FLRG/Fstl3 in Mouse samples. It is validated for Western Blot.Specifications
Follistatin-related Gene Protein/FLRG/Fstl3 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q9EQC7 | |
10272 | |
Mouse myeloma cell line NS0-derived recombinant mouse FLRG Val24-Val256 Accession # Q9EQC7 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse FLRG in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows 50-100% cross-reactivity with recombinant human FLRG. |
210725 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLRGFSRP, Follistatin-like 3, follistatin-like 3 (secreted glycoprotein), Follistatin-like protein 3, Follistatin-related gene protein, follistatin-related protein 3, FSTL3 | |
FSTL3 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only