missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse MIP-1 gamma (CCL9/10) Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 250-12-500UG
This item is not returnable.
View return policy
Description
MIP-1gamma is a CC chemokine found in murine blood and a wide variety of murine tissues, with no known human homolog. MIP-1gamma signals through the CCR1 receptor. MIP-1gamma chemoattracts neutrophils, and also inhibits colony formation of bone marrow myeloid immature progenitors. MIP-1gamma contains six cysteines, including the four highly conserved cysteine residues present in CC chemokines.Specifications
P51670 | |
protein with no preservative | |
11.6 | |
500 μg | |
-20°C | |
<1 EU/μg | |
Ccl9 | |
Unconjugated | |
QITHATETKE VQSSLKAQQG LEIEMFHMGF QDSSDCCLSY NSRIQCSRFI GYFPTSGGCT RPGIIFISKR GFQVCANPSD RRVQRCIERL EQNSQPRTYK Q | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
ELISA, Functional Assay, Western Blot | |
20308 | |
Mouse MIP-1 gamma (CCL9/10) | |
E. coli | |
RUO | |
C-C motif chemokine 9; CCF18; Ccl9; CCL9(29-101); CCL9(30-101); CCL9(31-101); chemokine (C-C motif) ligand 9; chemokine CCL9; macrophage inflammatory protein 1-gamma; Macrophage inflammatory protein-related protein 2; MIP-1 gamma; MIP-1-gamma; Mrp2; MRP-2; RP23-320E6.3; Scya10; Scya9; small inducible cytokine A10; small inducible cytokine A9; Small-inducible cytokine A9 | |
Mouse | |
Recombinant | |
Lyophilized |