missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse MIP-1 gamma (CCL9/10) Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£65.00 - £2410.00
Specifications
Accession Number | P51670 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 20308 |
Molecular Weight (g/mol) | 11.6 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17871493
|
Gibco™
250-12-100UG |
100 μg |
£482.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17801023
|
Gibco™
250-12-1MG |
1 mg |
£2410.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17840743
|
Gibco™
250-12-20UG |
20 μg |
£158.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17831883
|
Gibco™
250-12-250UG |
250 μg |
£842.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17850733
|
Gibco™
250-12-500UG |
500 μg |
£1442.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17811733
|
Gibco™
250-12-5UG |
5 μg |
£65.00
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MIP-1gamma is a CC chemokine found in murine blood and a wide variety of murine tissues, with no known human homolog. MIP-1gamma signals through the CCR1 receptor. MIP-1gamma chemoattracts neutrophils, and also inhibits colony formation of bone marrow myeloid immature progenitors. MIP-1gamma contains six cysteines, including the four highly conserved cysteine residues present in CC chemokines.Specifications
P51670 | |
protein with no preservative | |
11.6 | |
E. coli | |
RUO | |
C-C motif chemokine 9; CCF18; Ccl9; CCL9(29-101); CCL9(30-101); CCL9(31-101); chemokine (C-C motif) ligand 9; chemokine CCL9; macrophage inflammatory protein 1-gamma; Macrophage inflammatory protein-related protein 2; MIP-1 gamma; MIP-1-gamma; Mrp2; MRP-2; RP23-320E6.3; Scya10; Scya9; small inducible cytokine A10; small inducible cytokine A9; Small-inducible cytokine A9 | |
Mouse | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
20308 | |
Mouse MIP-1 gamma (CCL9/10) | |
-20°C | |
<1 EU/μg | |
Ccl9 | |
Unconjugated | |
QITHATETKE VQSSLKAQQG LEIEMFHMGF QDSSDCCLSY NSRIQCSRFI GYFPTSGGCT RPGIIFISKR GFQVCANPSD RRVQRCIERL EQNSQPRTYK Q | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |