missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse RELM alpha Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 450-26-1MG
This item is not returnable.
View return policy
Description
RELM alpha, also known as Resistin-like alpha or FIZZ1, is a small cytokine (88 aminoacids, MW 9.4 kDa) produced mostly by macrophages. Its highest constitutive expression has been observed in small peritoneal macrophages, alveolar macrophages and adipose tissue macrophages. It is strongly induced by IL-4 and IL-13. Together with Arginase 1 and Ym-1, RELM alpha is considered one of the hallmarks of the M2 polarization of mouse macrophages, however, macrophages polarized with IL-10 (M2c) express neither RELM alpha nor Arginase 1. Also, the expression patterns of RELM alpha and Arginase 1 in untreated tissue macrophages significantly differ. Elevated levels of RELM alpha may indicate parasitic infection. Humans do not have RELM alpha gene but human Resistin resembles mouse RELM alpha in respect to sequence, tissue distribution and function. Other members of RELM family in mouse: RELM beta, RELM gamma and Resistin display different tissue distribution and functional properties. RELM alpha plays an important role in parasite infection, mucosal inflammation and allergy.Specifications
Q9EP95 | |
protein with no preservative | |
10 | |
1 mg | |
-20°C | |
<1 EU/μg | |
Retnla | |
Unconjugated | |
MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
ELISA, Functional Assay, Western Blot | |
57262 | |
Mouse RELM alpha | |
E. coli | |
RUO | |
1810019L16Rik; cysteine-rich secreted protein A12-gamma; Cysteine-rich secreted protein FIZZ1; Fizz1; Fizz-1; FIZZ2; found in inflammatory zone 1; Himf; HXCP2; Hypoxia-induced mitogenic factor; parasite-induced macrophage novel gene 1 protein; Pmng1; RELMa; RELMalpha; RELM-alpha; RELMb; RELMbeta; RELM-beta; resistin like alpha; resistin-like alpha; Retnla; Xcp2 | |
Mouse | |
Recombinant | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ Mouse RELM alpha Recombinant Protein, PeproTech® > 1 mg
Spot an opportunity for improvement?Share a Content Correction