Learn More
Novus Biologicals™ Mouth Tissue Slides (Squamous Cell Carcinoma)- Paraffin
Brand: Novus Biologicals™ NBP2-30287
132.80 GBP valid until 2025-03-28
Use promo code "25306" to get your promotional price.
Description
These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.
Specifications
Human | |
Immunohistochemistry-Paraffin | |
Human |
Mouth Tissue Slides (Squamous Cell Carcinoma) | |
This product is for research use only and is not approved for use in humans or in clinical diagnosis. | |
Squamous Cell Carcinoma |
Product Suggestions
Customers who viewed this item also viewed.